Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPINK7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557601 Supplier Invitrogen™ Supplier No. PA557601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITYGNECHLC TESLKSNGRV QFLHDG Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

SPINK7 (serine peptidase inhibitor, Kazal type 7), also known as Ecg2 (esophagus cancer-related gene 2 protein), is a 76 amino acid secreted protein. Containing one Kazal-like domain, SPINK7 is thought to be a serine protease inhibitor. The gene that encodes SPINK7 maps to human chromosome 5, which contains 181 million base pairs encoding around 1,000 genes. Chromosome 5 is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.Probable serine protease inhibitor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPINK7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPINK7
Gene Accession No. P58062
Gene Alias ECG2; ECRG2; ECRG-2; EG408198; esophagus cancer related gene 2; esophagus cancer-related gene 2 protein; esophagus cancer-related gene-2; esophagus cancer-related protein 2; serine peptidase inhibitor, Kazal type 7; serine peptidase inhibitor, Kazal type 7 (putative); serine protease inhibitor Kazal-type 7; SPINK7; UNQ745/PRO1474
Gene Symbols SPINK7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPINK7. Recombinant protein control fragment (Product #RP-96888).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84651
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.