Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPINK9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558324
Description
Immunogen sequence: KQMVDCSHYK KLPPGQQRFC HHMYDPICGS DGKTYKNDCF FCSKVKKTDG TLKFVH Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 38%.
SPINK9 is a serine protease inhibitor which specifically inhibits KLK5. It may contribute to the regulation of the desquamation process in skin by inhibiting KLK5.
Specifications
| SPINK9 | |
| Polyclonal | |
| Unconjugated | |
| SPINK9 | |
| LEKTI2; lymphoepithelial Kazal-type-related inhibitor 2; serine peptidase inhibitor, Kazal type 9; serine protease inhibitor Kazal-type 9; SPINK9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 643394 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5DT21 | |
| SPINK9 | |
| Recombinant protein corresponding to Human SPINK9. Recombinant protein control fragment (Product #RP-96983). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction