Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPINLW1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537403 Supplier Invitrogen™ Supplier No. PA537403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 15 kDa. Purity is >95% by SDS-PAGE.

Eppin, an epididymal protease inhibitor, belongs to the WFDC family and to the telomeric cluster. The gene encoding the protein is localized to chromosome 20q12-q13 in centromeric and telomeric clusters. It expresses three mRNAs encoding two isoforms of a cystine-rich protein that contains Kunitz-type and WAP-type (four disulfide core) protease inhibitor consensus sequences. The mouse gene lies in a cluster of putative Eppin-like genes on mouse chromosome 2. Following ejaculation, Eppin is bound to semenogelin in seminal plasma and on human spermatozoa. This complex of Eppin and Semenogelin can provide antimicrobial activity for spermatozoa. It can also provide for the preparation and survival of spermatozoa for fertility in the female reproductive tract. Eppin, which is a secreted protein, is expressed in epididymis and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPINLW1
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene EPPIN
Gene Accession No. D4A2Z2, O95925, Q9DA01
Gene Alias 1700024E17Rik; cancer/testis antigen 71; cancer/testis antigen 72; CT71; CT72; dJ461P17.2; epididymal peptidase inhibitor; epididymal protease inhibitor; EPPIN; EPPIN1; EPPIN2; EPPIN3; protease inhibitor WAP7; serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin); serine protease inhibitor-like with Kunitz and WAP domains 1; serine protease inhibitor-like, with Kunitz and WAP domains 1 (eppin); Spinlw1; WAP four-disulfide core domain protein 7; WAP four-disulfide core domain protein 8; WAP7; WFDC7; Wfdc8
Gene Symbols EPPIN
Host Species Rabbit
Immunogen Recombinant full length human SPINLW1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57119, 685161, 75526
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.