Learn More
Invitrogen™ SPINLW1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total EPPIN protein.
Specifications
Specifications
| Antigen | SPINLW1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | EPPIN |
| Gene Accession No. | O95925, Q9DA01 |
| Gene Alias | 1700024E17Rik; cancer/testis antigen 71; cancer/testis antigen 72; CT71; CT72; dJ461P17.2; epididymal peptidase inhibitor; epididymal protease inhibitor; EPPIN; EPPIN1; EPPIN2; EPPIN3; protease inhibitor WAP7; serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin); serine protease inhibitor-like with Kunitz and WAP domains 1; serine protease inhibitor-like, with Kunitz and WAP domains 1 (eppin); Spinlw1; WAP four-disulfide core domain protein 7; WAP four-disulfide core domain protein 8; WAP7; WFDC7; Wfdc8 |
| Gene Symbols | EPPIN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.