Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spinophilin/Neurabin 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981981
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1981 Supplier Bioss Supplier No. BS12146R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

interacts with protein phosphatase 1 and actin filaments; involved in regulation of synaptic transmission.

Specifications

Antigen Spinophilin/Neurabin 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ppp1r9b
Gene Accession No. Q96SB3
Gene Alias Neb2; neurabin 2; neurabin II; Neurabin-2; Neurabin-II; Neural tissue-specific F-actin-binding protein II; nuerabin 2; p130; PP1bp134; PPP1R6; PPP1R9; Ppp1r9b; Protein phosphatase 1 regulatory subunit 9B; protein phosphatase 1, regulatory (inhibitor) subunit 9B; protein phosphatase 1, regulatory subunit 9B; protein phosphatase 1, regulatory subunit 9B, spinophilin; Spino; Spinophilin; SPL; Spn
Gene Symbols Ppp1r9b
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Spinophilin/Neurabin 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84687
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.