Learn More
SPINT1 Mouse anti-Human, Clone: OTI4E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504562

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.Specifications
SPINT1 | |
Monoclonal | |
Unconjugated | |
SPINT1 | |
HAI, HAI1, MANSC2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6692 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43278 | |
SPINT1 | |
Full length human recombit protein of human SPINT1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.