Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPINT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582939 Supplier Invitrogen™ Supplier No. PA582939
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTIKDLLPNV CAFPMEKGPC QTYMTRWFFN FETGECELFA YGGCGGNSNN FLRKEKCEKF CKFT Highest antigen sequence indentity to the following orthologs: Rat - 42%, Mouse - 44%.

SPINT3 is a protein coding gene. Gene ontology (GO) annotation include extracellular region/cellular component.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPINT3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPINT3
Gene Accession No. P49223
Gene Alias HKIB9; Kunitz-type protease inhibitor 3; serine peptidase inhibitor, Kunitz type 3; serine peptidase inhibitor, Kunitz type, 3; serine protease inhibitor Kunitz type 3; serine protease inhibitor, Kunitz type, 3; SPINT3
Gene Symbols SPINT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPINT3. Recombinant protein control fragment (Product #RP-108189).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10816
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.