Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPINT4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583834
Description
Immunogen sequence: KICGDLKDPC KLDMNFGSCY EVHFRYFYNR TSKRCETFVF SGCNGNLNNF KLK.
SPINT4 is a protein coding gene. Gene ontology (GO) annotation include extracellular region/cellular component.
Specifications
SPINT4 | |
Polyclonal | |
Unconjugated | |
SPINT4 | |
C20orf137; dJ601O1.1; Kunitz-type protease inhibitor 4; serine peptidase inhibitor, Kunitz type 4; serine protease inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 4; SPINT3; SPINT4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
391253 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UDR6 | |
SPINT4 | |
Recombinant protein corresponding to Human SPINT4. Recombinant protein control fragment (Product #RP-108198). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction