Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spiramycin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175409
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175409 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175409 Supplier Invitrogen™ Supplier No. PA175409
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 2.5 μg/mL. Sensitivity: 10 ng/mL Spiramycin produces 77% inhibition in a competitive ELISA employing Spiramycin polyclonal antibody.

Aminoglycosides are a family of bacterial antibiotics that are used in the treatment of specific bacterial infections. They display a concentration dependent killing action and are active against a wide range of aerobic Gram-negative bacilli. Aminoglycosides are molecules that are comprised of an amino group and a sugar group. They operate by inhibiting the bacteria from producing proteins vital to its growth. More specifically, they bind to the bacterial 30S ribosomal subunit where they prevent the translocation of the peptidyl-tRNA from the A-site to the P-site, subsequently giving rise to a misreading of mRNA resulting in the inhibition of protein synthesis. This consequently results in a disruption to the integrity of the bacterial cell membrane . In addition to their use to prevent bacterial infection, aminoglycosides have been used as growth promoters in food producing animals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spiramycin
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias Rovamycine
Host Species Sheep
Immunogen Spiramycin-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.