Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPIRE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559362
Description
Immunogen sequence: LQRGESSMRS EKPSTAHHRP LRSIARFSSK SKSMDKSDEE LQFPKELMED WSTMEVCVDC KKFISEIISS SRRSLVLANK RARLKRKTQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Acts as a actin nucleation factor, remains associated with the slow-growing pointed end of the new filament. Involved in intracellular vesicle transport along actin fibers, providing a novel link between actin cytoskeleton dynamics and intracellular transport. Required for asymmetric spindle positioning and asymmetric cell division during meiosis. Required for normal formation of the cleavage furrow and for polar body extrusion during female germ cell meiosis.
Specifications
SPIRE1 | |
Polyclonal | |
Unconjugated | |
SPIRE1 | |
KIAA1135; Protein spire homolog 1; SPIR1; Spir-1; spire actin nucleation factor 1; spire family actin nucleation factor 1; spire homolog 1; spire type actin nucleation factor 1; SPIRE1; spire-type actin nucleation factor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56907 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q08AE8 | |
SPIRE1 | |
Recombinant protein corresponding to Human SPIRE1. Recombinant protein control fragment (Product #RP-98282). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction