Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spironolactone, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18606977 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-977 25 g Amber Glass Bottle
18-606-976 5 g Amber Glass Bottle
18-606-979 100 g Amber Glass Bottle
18-606-978 500 g Amber Glass Bottle
18-606-980 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-977 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SP32025GM
Add to Cart
Add to Cart

SP320, 52-01-7, C24H32O4S

Spectrum™ Chemical Spironolactone, USP is used to treat high blood pressure and edema. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 52-01-7
Molecular Formula C24H32O4S
Molecular Weight (g/mol) 416.58
InChI Key LXMSZDCAJNLERA-ZHYRCANASA-N
IUPAC Name (1R,3aS,3bR,4R,9aR,9bS,11aS)-4-(acetylsulfanyl)-9a,11a-dimethyl-2,3,3a,3b,4,5,7,8,9,9a,9b,10,11,11a-tetradecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione
SMILES CC(=O)S[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]2CC[C@@]3(C)[C@@H](CC[C@@]33CCC(=O)O3)[C@H]12

Specifications

CAS Max % 1
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation -41° to -45°
Percent Purity 97 to 103%
Grade USP
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.