CAS RN 52-01-7
Thermo Scientific Chemicals Spironolactone, 99%
CAS: 52-01-7 Molecular Formula: C24H32O4S Molecular Weight (g/mol): 416.57 MDL Number: MFCD00082250 InChI Key: LXMSZDCAJNLERA-ZHYRCANASA-N Synonym: spironolactone,aldactone,spirolactone,verospiron,euteberol,spiroctan,spirolang,verospirone,aldactone a,spironocompren PubChem CID: 5833 ChEBI: CHEBI:9241 IUPAC Name: S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate SMILES: CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
Spironolactone
CAS: 52-01-7 Molecular Formula: C24H32O4S Molecular Weight (g/mol): 416.576 MDL Number: MFCD00082250 InChI Key: LXMSZDCAJNLERA-ZHYRCANASA-N PubChem CID: 5833 ChEBI: CHEBI:9241 IUPAC Name: S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate SMILES: CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
Spironolactone, USP, 97-103%, Spectrum™ Chemical
CAS: 52-01-7 Molecular Formula: C24H32O4S Molecular Weight (g/mol): 416.58 InChI Key: LXMSZDCAJNLERA-ZHYRCANASA-N IUPAC Name: (1R,3aS,3bR,4R,9aR,9bS,11aS)-4-(acetylsulfanyl)-9a,11a-dimethyl-2,3,3a,3b,4,5,7,8,9,9a,9b,10,11,11a-tetradecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione SMILES: CC(=O)S[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]2CC[C@@]3(C)[C@@H](CC[C@@]33CCC(=O)O3)[C@H]12
Spironolactone, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.
Spironolactone 98.0+%, TCI America™
CAS: 52-01-7 Molecular Formula: C24H32O4S Molecular Weight (g/mol): 416.576 MDL Number: MFCD00082250 InChI Key: LXMSZDCAJNLERA-ZHYRCANASA-N Synonym: spironolactone,aldactone,spirolactone,verospiron,euteberol,spiroctan,spirolang,verospirone,aldactone a,spironocompren PubChem CID: 5833 ChEBI: CHEBI:9241 IUPAC Name: S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate SMILES: CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
R&D Systems™ Spironolactone
CAS: 52-01-7 Molecular Formula: C24H32O4S