Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Splicing Factor 1 Antibody (OTI7H9), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274338JF549
Description
Splicing Factor 1 Monoclonal specifically detects Splicing Factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Splicing Factor 1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SF1 | |
Recombinant protein fragment corresponding to amino acids 1-276 of human Splicing Factor 1 (NP_973727) produced in E.coli. | |
0.1 mL | |
7536.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI7H9 | |
Western Blot, Immunohistochemistry | |
Mammalian branch point-binding protein, mBBP, splicing factor 1, Transcription factor ZFM1, ZFM1BBP, Zinc finger gene in MEN1 locus, Zinc finger protein 162D11S636, ZNF162MBBP | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction