Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Splicing factor, arginine/serine-rich 11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Splicing factor, arginine/serine-rich 11 |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Splicing factor, arginine/serine-rich 11 Polyclonal specifically detects Splicing factor, arginine/serine-rich 11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Splicing factor, arginine/serine-rich 11 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9295 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LLPTPNPLTQIGAVPLAALGAPTLDPALAALGLPGANLNSQSLAADQLLKLMSTVDPKLNHVAAGLVSPSLKSDTSSKEIEEAMKRVREAQSLISAAIEPDKKEEKRRHSRSRSR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
Arginine-rich 54 kDa nuclear protein, dJ677H15.2, DKFZp686M13204, p54NET2, serine/arginine-rich splicing factor 11, SFRS11, splicing factor p54, Splicing factor, arginine/serine-rich 11FLJ18226, SR splicing factor 11 | |
SRSF11 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title