Learn More
Therapak™ Split Collection Kit
Catalog No. 22131556 Shop All Therapak Corporation Products
Change view
Click to view available options
Product Type:
Split Collection Bag Kit
Split Collection Box Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-131-556 Split Collection Bag Kit
22-131-557 Split Collection Box Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-131-556 Supplier Therapak™ Supplier No. 40002
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Kit includes one wide-mouth 6.5 oz. collection cup with pour spout and temperature strip, two sterile 60mL containers with sterility seal, and split transport bag with absorbent; with or without transport box.
For drugs of abuse testing
Designed for testing applications requiring two samples from a single donor.
Specifications
Product Type Split Collection Bag Kit
For Use With (Application) For drugs of abuse testing
Includes 6.5 oz. Beaker style collection cup with pour-spout and temperature strip, two 60 mL sterile containers with sterility seal and a transport bag with absorbent