Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Siltek™ Liner
SDP

Catalog No. 06717816 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Includes:
Siltek Cup Splitter
Siltek Cup Splitter w/CarboFrit
Siltek Cup Splitter w/Wool
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
06-717-816 Siltek Cup Splitter w/CarboFrit
06-717-813 Siltek Cup Splitter w/Wool
06-717-814 Siltek Cup Splitter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-717-816 Supplier Restek™ Supplier No. 20710216
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For high or low molecular weight compounds

  • Increased sample residence time in the liner for homogeneous vaporization
  • 4mm I.D.
  • With Siltek™ deactivation
  • Available with or without packing

Specifications

Product Type Siltek Cup Splitter Inlet Liner
Packing Material CarboFrit
Length (Metric) 7.85 cm
Quantity 5/Pk.
Includes Siltek Cup Splitter w/CarboFrit
Diameter (Metric) Inner 0.4 cm
Diameter (Metric) Outer 0.63 cm
For Use With (Equipment) Agilent GCs Equipped with Split/Splitless Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.