Learn More
Trajan™ CTC PAL Fixed Needle Syringe
SDP
Catalog No. p-8014049 Shop All SGE Inc. Products
Change view
Click to view available options
Needle Gauge:
22 G
Diameter (Metric) Outer Needle:
0.72 mm
Needle Point Style:
Side Hole
Catalog No. Needle Gauge Diameter (Metric) Outer Needle Needle Point Style
03-350-178 22 G 0.72 mm Side Hole
1 options
Catalog No. 03-350-178 Supplier Trajan™ Supplier No. 002983
Borosilicate glass barrel and stainless steel needles provide strength and inertness. Backing provides quick and accurate volume assessment.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
Specifications
Description Gas tight
Needle Gauge 22 G
Diameter (Metric) Outer 0.72 mm
Diameter (Metric) Outer Needle 0.72 mm
Product Type Fixed Needle Syringe
Model 10F-CTC-GT-5/0.72H
Quantity 1 Ea.
Volume (Metric) 10 μL
Needle Point Style Side Hole