Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Splunc2 Monoclonal Antibody (4C7D7)
GREENER_CHOICE

Catalog No. PIMA517094
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517094 Supplier Invitrogen™ Supplier No. MA517094
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17094 targets Splunc2 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17094 immunogen is purified recombinant fragment of human Splunc2 (amino acids: 16-250) expressed in E. Coli. MA5-17094 detects Splunc2 which has a predicted molecular weight of approximately 27kDa.

SPLUNC2 is one member of PLUNC family, the gene undergoes alternative splicing using two 5' non-coding exons, suggesting that the gene is regulated by alternative promoters.multiple SPLUNC2 isoforms are found in the oral cavity and suggest that these proteins may be differentially regulated in distinct tissues where they may function in the innate immune response. Mucin plugs, mucous and intermediate cells of mucoepidermoid carcinomas were positive for LPLUNC1 and SPLUNC2, but areas composed of epidermoid and clear cells were negative for all PLUNCs. Papillary cystadenocarcinoma was positive for all PLUNCs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Splunc2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4C7D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene BPIFA2
Gene Accession No. Q96DR5
Gene Alias bA49G10.1; BPI fold containing family A member 2; BPI fold containing family A, member 2; BPI fold containing family A, member 2E; BPI fold-containing family A member 2; BPIFA2; Bpifa2e; C20orf70; mSplunc2; parotid secretory protein; PSP; short palate; short palate, lung and nasal epithelium carcinoma-associated protein 2; SPLUNC2; UNQ510/PRO1025
Gene Symbols BPIFA2
Host Species Mouse
Immunogen Purified recombinant fragment of human Splunc2 (amino acids: 16-249) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140683
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.