Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Polyacrylate (PA), df 85 μm
Shop All MilliporeSigma Supelco Products
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
Needle Gauge:
23 ga
24 ga
Specifications
Specifications
| For Use With (Application) | SPME |
| For Use With (Equipment) | Autosampler |
| Type | SPME Fibers and Accessories |
| Material | Fused Silica Fiber |
| Needle Gauge | 23 ga |
| Quantity | 3/Pk. |