Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm, StableFlex™
Solid phase microextraction technique can be used for GC/MS analysis. It helps in simplifying and speeds up the sample preparation. It also reduces solvent requirement for the extraction.
$818.00 - $1080.00
Specifications
For Use With (Application) SPME
Type SPME Fibers and Accessories
Quantity 3/Pk.
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Material Needle Gauge Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Material Needle Gauge Price Quantity & Availability  
11-102-0744
View Documents
MilliporeSigma™ Supelco™
57293U
Autosampler Polydimethylsiloxane/Divinylbenzene 23 ga
Each for $1,080.00
Only null left
Add to Cart
11-100-8572
View Documents
MilliporeSigma™ Supelco™
57327U
Autosampler Plain pink hub 24 ga
Each for $818.00
Only null left
Add to Cart
11-100-8486
View Documents
MilliporeSigma™ Supelco™
57346U
Manual Holder Fused Silica Fiber 23 ga
Each for $915.00
Only null left
Add to Cart
11-100-8582
View Documents
MilliporeSigma™ Supelco™
57326U
Manual Holder Plain pink hub 24 ga
Each for $838.00
Only null left
Add to Cart
Description
Description
Specifications
Specifications
SPME
SPME Fibers and Accessories
3/Pk.
Videos
SDS
Documents
Documents
Product Certifications
StableFlex is a trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.