Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Polydimethylsiloxane (PDMS), df 100 μm
Catalog No. p-7194119 Shop All MilliporeSigma Supelco Products
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
Material:
Fused Silica Fiber
Metal Alloy Fiber
Needle Gauge:
23 ga
24 ga
Catalog No. 11-102-0750
Supplier: MilliporeSigma™ Supelco™ 57341U
Only null left
Add to Cart
Add to Cart
Solid phase microextraction technique can be used for GC/MS analysis. It helps in simplifying and speeds up the sample preparation. It also reduces solvent requirement for the extraction.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Fused Silica Fiber
Needle Gauge 23 ga
Quantity 3/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.