Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assortment Kit 1
Catalog No. p-7194122 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Manual Holder
Needle Gauge:
24 ga
Catalog No. For Use With (Equipment) Needle Gauge
11-100-8584 Manual Holder 24 ga
1 options
Catalog No. 11-100-8584 Supplier MilliporeSigma™ Supelco™ Supplier No. 57306
Only null left
Add to Cart
Add to Cart
Kit 1 - For Volatiles and Semivolatiles - contains one fiber of each: 85 μm polyacrylate coating, 100 μm polydimethylsiloxane coating and 7 μm polydimethylsiloxane coating
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Manual Holder
Type SPME Fibers and Accessories
Material Fused Silica Fiber
Needle Gauge 24 ga