Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assortment Kit 4
Catalog No. p-7194166 Shop All MilliporeSigma Supelco Products
Click to view available options
For Use With (Equipment):
Autosampler
Catalog No. 11-100-8238
Supplier: MilliporeSigma™ Supelco™ 57287U
Only null left
Add to Cart
Add to Cart
Kit 4 - For Flavors and Odors - contains one of each: 100 μm polydimethylsiloxane coating, 65 μm polydimethylsiloxane/divinylbenzene coating and 75 μm Carboxen/polydimethylsiloxane coating
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Fused Silica Fiber
Needle Gauge 23 ga
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.