Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPNS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560188 Supplier Invitrogen™ Supplier No. PA560188
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RAQLHVQGLL HEAGSTDDRI VVPQRGRSTR VPVASVLI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 84%.

SPINSTER, also known as SPNS1 or SPIN1, is a 528 amino acid multi-pass membrane protein that localizes to the inner mitochondrial membrane and belongs to the spinster subfamily of the major facilitator superfamily. SPINSTER interacts with Bcl-x and Bcl-2 and, via this interaction, is thought to be involved in necrotic or autophagic cell death. The related protein SPNS2 is critical for the normal lymphocyte localization and mammalian immune system function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPNS1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPNS1
Gene Accession No. Q9H2V7
Gene Alias HSpin1; LAT; nrs; PP2030; PP20300; Protein spinster homolog 1; sphingolipid transporter 1 (putative); SPIN1; SPINL; spinster homolog 1; spinster-like protein 1; SPNS sphingolipid transporter 1 (putative); SPNS1
Gene Symbols SPNS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPNS1. Recombinant protein control fragment (Product #RP-98580).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.