Learn More
Invitrogen™ SPNS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113920
Description
Immunogen sequence: AGSDTAPFLS QADDPDDGPV PGTPGLPGST GNPKSEEPEV PDQEGLQRIT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 91%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
SPINSTER, also known as SPNS1 or SPIN1, is a 528 amino acid multi-pass membrane protein that localizes to the inner mitochondrial membrane and belongs to the spinster subfamily of the major facilitator superfamily. SPINSTER interacts with Bcl-x and Bcl-2 and, via this interaction, is thought to be involved in necrotic or autophagic cell death. The related protein SPNS2 is critical for the normal lymphocyte localization and mammalian immune system function.
Specifications
| SPNS1 | |
| Polyclonal | |
| Unconjugated | |
| SPNS1 | |
| HSpin1; LAT; nrs; PP2030; PP20300; Protein spinster homolog 1; sphingolipid transporter 1 (putative); SPIN1; SPINL; spinster homolog 1; spinster-like protein 1; SPNS sphingolipid transporter 1 (putative); SPNS1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 83985 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H2V7 | |
| SPNS1 | |
| Synthetic peptide directed towards the N terminal region of human SPNS1 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.