Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPNS2 Rabbit anti-Drosophila, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502209230
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9230 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9230 Supplier Fabgennix Inc Supplier No. SPNS2101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPNS2 Polyclonal antibody specifically detects SPNS2 in Drosophila, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen SPNS2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene SPNS2
Gene Accession No. Q91VM4
Gene Alias Protein spinster homolog 2; sphingolipid transporter 2; SPNS2
Host Species Rabbit
Immunogen Synthetic peptide taken within TMD9 and TMD10 on human SPNS2 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100270678, 216892, 748255
Target Species Drosophila, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.