Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPNS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548420
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548420 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548420 Supplier Invitrogen™ Supplier No. PA548420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPNS3 Polyclonal Antibody for IHC (P), Western Blot

Sphingolipid transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPNS3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SPNS3
Gene Accession No. Q6ZMD2
Gene Alias Protein spinster homolog 3; sphingolipid transporter 3 (putative); spinster homolog 3; SPNS sphingolipid transporter 3 (putative); SPNS3
Gene Symbols SPNS3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 470-498 amino acids from the C-terminal region of human SPNS3.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201305
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.