Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPO11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530549 Supplier Invitrogen™ Supplier No. PA530549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (83%), Rat (85%), Pig (91%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Meiotic recombination and chromosome segregation require the formation of double-strand breaks (DSBs) in paired chromosome homologs. During meiosis in yeast, a meiotic recombination protein is covalently-linked to the 5' end of DSBs and is essential for the formation of DSBs. The protein encoded by this gene is similar in sequence and conserved features to the yeast meiotic recombination protein. The encoded protein belongs to the TOP6A protein family. Several transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPO11
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SPO11
Gene Accession No. Q9Y5K1
Gene Alias AI449549; Cancer/testis antigen 35; CT35; hypothetical protein LOC402982; meiotic recombination protein SPO11; MGC39953; SPATA43; spermatogenesis associated 43; SPO11; SPO11 initiator of meiotic double stranded breaks; SPO11 meiotic protein covalently bound to DSB; SPO11 meiotic protein covalently bound to DSB homolog; SPO11 meiotic protein covalently bound to DSB homolog (S. cerevisiae); SPO11, initiator of meiotic double stranded breaks; sporulation protein, meiosis-specific, SPO11 homolog; sporulation protein, meiosis-specific, SPO11 homolog (S. cervisiae); TOPVIA; zgc:77876
Gene Symbols SPO11
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SPO11. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23626
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.