Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPO11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544237
Description
Peptide sequence: KFSLILKILS MIYKLVQSNT YATKRDIYYT DSQLFGNQTV VDNIINDISC Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%; Zebrafish: 77%.
Meiotic recombination and chromosome segregation require the formation of double-strand breaks (DSBs) in paired chromosome homologs. During meiosis in yeast, a meiotic recombination protein is covalently-linked to the 5' end of DSBs and is essential for the formation of DSBs. The protein encoded by this gene is similar in sequence and conserved features to the yeast meiotic recombination protein. The encoded protein belongs to the TOP6A protein family. Several transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been described.
Specifications
| SPO11 | |
| Polyclonal | |
| Unconjugated | |
| SPO11 | |
| AI449549; Cancer/testis antigen 35; CT35; hypothetical protein LOC402982; meiotic recombination protein SPO11; MGC39953; SPATA43; spermatogenesis associated 43; SPO11; SPO11 initiator of meiotic double stranded breaks; SPO11 meiotic protein covalently bound to DSB; SPO11 meiotic protein covalently bound to DSB homolog; SPO11 meiotic protein covalently bound to DSB homolog (S. cerevisiae); SPO11, initiator of meiotic double stranded breaks; sporulation protein, meiosis-specific, SPO11 homolog; sporulation protein, meiosis-specific, SPO11 homolog (S. cervisiae); TOPVIA; zgc:77876 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23626 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y5K1 | |
| SPO11 | |
| Synthetic peptide directed towards the N-terminal of human SPO11 (aa 110-159). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction