Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPO11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142463
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142463 Supplier Thermo Scientific Supplier No. PA5142463
Add to Cart
Add to Cart
This item is not returnable. View return policy

SPO11 Polyclonal Antibody

SPO11 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SPO11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SPO11
Gene Accession No. Q9Y5K1
Gene Alias AI449549; Cancer/testis antigen 35; CT35; hypothetical protein LOC402982; meiotic recombination protein SPO11; MGC39953; SPATA43; spermatogenesis associated 43; SPO11; SPO11 initiator of meiotic double stranded breaks; SPO11 meiotic protein covalently bound to DSB; SPO11 meiotic protein covalently bound to DSB homolog; SPO11 meiotic protein covalently bound to DSB homolog (S. cerevisiae); SPO11, initiator of meiotic double stranded breaks; sporulation protein, meiosis-specific, SPO11 homolog; sporulation protein, meiosis-specific, SPO11 homolog (S. cervisiae); TOPVIA; zgc:77876
Gene Symbols SPO11
Host Species Goat
Immunogen Peptide with sequence C-NTYATKRDIYYTDSQ (aa 128-142).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23626
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.