Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPOCK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598497
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598497 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598497 Supplier Invitrogen™ Supplier No. PA598497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPOCK1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

SPOCK1, also known as Testican-1, is a gene located on chromosome 5q31.2, encoding a member of the proteoglycan family characterized by containing multiple domains that facilitate interactions with other extracellular matrix proteins. SPOCK1 is primarily expressed in various tissues, including the brain and reproductive organs, where it plays crucial roles in modulating cell adhesion, migration, and matrix remodeling. Functionally, SPOCK1 contributes to the regulation of extracellular matrix composition, influencing processes such as tissue development and wound healing. In the context of cancer, SPOCK1 is often upregulated, promoting tumor progression and metastasis by enhancing cancer cell motility and invasiveness. Its involvement in these pathological mechanisms makes SPOCK1 a potential biomarker for cancer prognosis and a target for therapeutic intervention. Research continues to unravel SPOCK1's diverse biological activities and its impact on cellular architecture and disease progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPOCK1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SPOCK1
Gene Accession No. Q08629, Q62288
Gene Alias FLJ37170; Protein SPOCK; sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1; sparc/osteonectin, cwcv and kazal-like domains proteoglycan 1; Spock; Spock1; testican; testican 1; testican-1; TIC1; TICN1
Gene Symbols SPOCK1
Host Species Rabbit
Immunogen Recombinant Human Testican-1 protein (290-435AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20745, 6695
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.