Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPOCK2 Monoclonal Antibody (327403)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524053

Catalog No. PIMA524053


Only null left
Add to Cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombinant human Testican 1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

The SPOCK2 gene encodes the protein testican-2, a member of the SPARC (secreted protein acidic and rich in cysteine) family of extracellular matrix (ECM) proteins. SPOCK2 is involved in modulating the ECM environment, influencing cell-matrix interactions, tissue remodeling, and cellular communication. It plays a role in the regulation of proteolytic activity by inhibiting metalloproteinases, thereby contributing to the maintenance and integrity of the ECM. SPOCK2 is prominently expressed in the brain and testes, highlighting its involvement in neural development and reproductive biology. In neural tissues, it influences processes such as neuronal differentiation, synaptogenesis, and axonal guidance, which are critical for proper brain development and function. Dysregulation of SPOCK2 expression is associated with pathological conditions, including cancer, where it may impact tumor progression and metastasis through alterations in cell adhesion and migration. Research into SPOCK2's specific interactions and mechanisms continues, providing insights into its potential as a therapeutic target for managing ECM-related disorders and diseases involving abnormal cell-matrix interactions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPOCK2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q92563
SPOCK2
Mouse myeloma cell line NS0-derived recombinant human Testican 2/SPOCK2 Glu23-Trp424.
100 μg
Primary
Human
Antibody
IgG1
Immunoprecipitation, Western Blot
327403
Unconjugated
SPOCK2
AA407235; FLJ97039; Gcap26; granule cell antiserum positive 26; KIAA0275; KIAA0275CWCV; mKIAA0275; sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2; sparc/osteonectin, cwcv and kazal-like domains proteoglycan 2; SPARC/osteonectin, CWCV, and Kazal-like domains proteoglycan 2; Spock2; Testican-2; TICN2; UNQ269/PRO306
Mouse
Protein A/G
RUO
9806
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.