Learn More
Invitrogen™ SPOCK3 Monoclonal Antibody (330404)

Description
In Western blots, approximately 30% cross-reactivity with recombinant human (rh) Testican 1 and rhTestican 3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | SPOCK3 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 330404 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Spock3 |
| Gene Accession No. | Q8BKV0 |
| Gene Alias | 2900045C01Rik; AI428471; HSAJ1454; mKIAA4039; sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3; sparc/osteonectin, cwcv and kazal-like domains proteoglycan 3; SPARC/osteonectin, CWCV, and Kazal-like domains proteoglycan 3; SPOCK3; TES-3; testican 3; testican-3; TICN3; UNQ409/PRO771 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.