Learn More
Invitrogen™ SPOCK3 Polyclonal Antibody

Description
Immunogen sequence: CPCPSDKPTS TSRNVKRACS DLEFREVANR LRDWFKALHE SGSQNKKTKT LLRPERSRFD TSILPICKDS LGWMFNRLDT NYDLLLDQSE LRSIYLDKNE QCTKAFFNSC DTYKDSLISN NEWCYCFQRQ QDPPCQTELS NIQKRQGVKK LLGQYIPLCD EDGYYKPTQC HGSVGQCWCV DRYGNEVMGS RINGVADCAI D.
Specifications
Specifications
| Antigen | SPOCK3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Spock3 |
| Gene Accession No. | Q8BKV0, Q9BQ16 |
| Gene Alias | 2900045C01Rik; AI428471; HSAJ1454; mKIAA4039; sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3; sparc/osteonectin, cwcv and kazal-like domains proteoglycan 3; SPARC/osteonectin, CWCV, and Kazal-like domains proteoglycan 3; SPOCK3; TES-3; testican 3; testican-3; TICN3; UNQ409/PRO771 |
| Gene Symbols | Spock3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.