Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPON1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7520R
Description
SPON1 is a cell adhesion protein that promotes the attachment of spinal cord and sensory neuron cells and the out growth of neurites in vitro. SPON1 may contribute to the growth and guidance of axons in both the spinal cord and the PNS. Is known to be a major factor for vascular smooth muscle cells.Specifications
| SPON1 | |
| Polyclonal | |
| Unconjugated | |
| SPON1 | |
| AI666765; AW455831; BC020531; D330035F22Rik; FSP; F-spondin; KIAA0762; MGC10724; Spon1; spondin 1; spondin 1, (f-spondin) extracellular matrix protein; spondin 1, extracellular matrix protein; Spondin-1; Sponf; Vascular smooth muscle cell growth-promoting factor; VSGP; VSGP/F-spondin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10418 | |
| -20°C | |
| Liquid |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| Q9HCB6 | |
| SPON1 | |
| KLH conjugated synthetic peptide derived from human SPON1/F spondin. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction