Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPON2 Polyclonal Antibody
SPON2 Polyclonal Antibody
Supplier: Thermo Scientific PA566822
Description
SPON2 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
SPON2 | |
Polyclonal | |
Unconjugated | |
SPON2 | |
2310045I24Rik; AI504350; differentially expressed in cancerous and non-cancerous lung cells 1; DIL1; DIL-1; DKFZp686G21139; FLJ16313; FLJ22401; FLJ34460; F-spondin; MINDIN; Mspondin; M-SPONDIN; SPON2; spondin 2; spondin 2, extracellular matrix protein; spondin-2; UNQ435/PRO866 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10417 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.15 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUD6 | |
SPON2 | |
Recombinant Human SPON2. Recombinant protein control fragment (Product #RP-105379). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction