Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPON2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113962
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113962 Supplier Invitrogen™ Supplier No. PA5113962
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSARAPAKYS ITFTGKWSQT AFPKQYPLFR PPAQWSSLLG AAHSSDYSMW For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

Cell adhesion protein that promotes adhesion and outgrowth of hippocampal embryonic neurons. Binds directly to bacteria and their components and functions as an opsonin for macrophage phagocytosis of bacteria. Essential in the initiation of the innate immune response and represents a unique pattern-recognition molecule in the ECM for microbial pathogens. Binds bacterial lipopolysaccharide (LPS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPON2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPON2
Gene Accession No. Q9BUD6
Gene Alias 2310045I24Rik; AI504350; differentially expressed in cancerous and non-cancerous lung cells 1; DIL1; DIL-1; DKFZp686G21139; FLJ16313; FLJ22401; FLJ34460; F-spondin; MINDIN; Mspondin; M-SPONDIN; SPON2; spondin 2; spondin 2, extracellular matrix protein; spondin-2; UNQ435/PRO866
Gene Symbols SPON2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human SPON2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10417
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.