Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPON2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA519294
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519294 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519294 Supplier Thermo Scientific Supplier No. PA519294

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

SPON2 Polyclonal antibody specifically detects SPON2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPON2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SPON2
Gene Accession No. Q9BUD6
Gene Alias 2310045I24Rik; AI504350; differentially expressed in cancerous and non-cancerous lung cells 1; DIL1; DIL-1; DKFZp686G21139; FLJ16313; FLJ22401; FLJ34460; F-spondin; MINDIN; Mspondin; M-SPONDIN; SPON2; spondin 2; spondin 2, extracellular matrix protein; spondin-2; UNQ435/PRO866
Gene Symbols SPON2
Host Species Goat
Immunogen Synthetic peptide sequence (RKNQYVSNGLRDFA) corresponding to the internal amino acids of SPON2
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10417
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.