Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPOP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116544
Description
Positive Samples: HeLa, HT-29 Immunogen sequence: ARSPVFSAMF EHEMEESKKN RVEINDVEPE VFKEMMCFIY TGKAPNLDKM ADDLLAAADK YALERLKVMC EDALCSNLSV ENAAEILILA DLHSADQLKT QAVDFINYHA SDVLETSGWK SMVVSHPHLV AEAYRSLASA QCPFLGPPRK RLKQS.
Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.
Specifications
SPOP | |
Polyclonal | |
Unconjugated | |
SPOP | |
AI315626; BTBD32; HIB homolog 1; Pcif1; PDX-1 C-terminal-interacting factor 1; roadkill homolog 1; similar to speckle-type POZ protein; speckle type BTB/POZ protein; speckle type POZ protein; speckle-type POZ protein; Spop; SPOP1; TEF2; zgc:64140 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8405 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.78 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O43791 | |
SPOP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 220-374 of human SPOP (NP_0010072271). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction