Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPOP Rabbit anti-Mouse, Zebrafish, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549673
Description
Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.
Specifications
SPOP | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6ZWS8, Q7T330 | |
SPOP | |
KLH conjugated synthetic peptide between 74-107 amino acids from the N-terminal region of human (DANRE) spop. | |
400 μL | |
Primary | |
Mouse, Zebrafish | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Spop | |
Speckle-type POZ protein, HIB homolog 1, SPOP1, spop | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100005514, 20747 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction