Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPOPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598171
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598171 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598171 Supplier Invitrogen™ Supplier No. PA598171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPOPL Polyclonal Antibody for Western Blot, ELISA

Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination and subsequent proteasomal degradation of target proteins, but with relatively low efficiency. Cullin-RING-based BCR (BTB-CUL3- RBX1) E3 ubiquitin-protein ligase complexes containing homodimeric SPOPL or the heterodimer formed by SPOP and SPOPL are less efficient than ubiquitin ligase complexes containing only SPOP. May function to down-regulate the activity of cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complexes that contain SPOP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPOPL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SPOPL
Gene Accession No. Q6IQ16
Gene Alias BTBD33; HIB homolog 2; roadkill homolog 2; speckle type BTB/POZ protein like; speckle type POZ protein like; speckle-type POZ protein-like; SPOPL
Gene Symbols SPOPL
Host Species Rabbit
Immunogen Recombinant Human Speckle-type POZ protein-like protein (1-120AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339745
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.