Learn More
Invitrogen™ SPPL2A Polyclonal Antibody

Description
Immunogen sequence: KFWKGNSYQMM DHLDCATNEE NPVISGEQIV QQ Highest antigen sequence identity to the following orthologs - mouse 79%, rat 73%.
Specifications
Specifications
| Antigen | SPPL2A |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SPPL2A |
| Gene Accession No. | Q8TCT8 |
| Gene Alias | 2010106G01Rik; C130089K23Rik; IMP3; IMP-3; intramembrane cleaving protease; intramembrane protease 3; MNCb-3763; Presenilin-like protein 2; protein SPPL2a; PSEC0147; PSL2; RGD1563001; signal peptide peptidase like 2A; signal peptide peptidase-like 2A; SPPL2A; SPPL2a protein; SPP-like 2A; SPP-like 2A protein |
| Gene Symbols | SPPL2A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.