Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPPL2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598784
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598784 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598784 Supplier Invitrogen™ Supplier No. PA598784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPPL2A Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

By searching sequence databases for homologs of Dictyostelium discoideum Impas, which shares homology with presenilin, followed by PCR of lymphocyte and hippocampus cDNA libraries, Grigorenko et al. (2002) cloned SPPL2a. This protein, which may act as an intramembrane protease, contains an N-terminal protease-associated (PA) domain, several transmembrane regions, a hydrophilic loop, conservative sequences around the first and second aspartate residues, and an invariant PAL motif near the C terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPPL2A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SPPL2A
Gene Accession No. Q8TCT8
Gene Alias 2010106G01Rik; C130089K23Rik; IMP3; IMP-3; intramembrane cleaving protease; intramembrane protease 3; MNCb-3763; Presenilin-like protein 2; protein SPPL2a; PSEC0147; PSL2; RGD1563001; signal peptide peptidase like 2A; signal peptide peptidase-like 2A; SPPL2A; SPPL2a protein; SPP-like 2A; SPP-like 2A protein
Gene Symbols SPPL2A
Host Species Rabbit
Immunogen Recombinant Human Signal peptide peptidase-like 2A protein (26-172AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84888
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.