Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPPL2B Polyclonal Antibody
SPPL2B Polyclonal Antibody
Supplier: Thermo Scientific PA542683
Description
SPPL2B Polyclonal Antibody for Western Blot, IHC

Specifications
SPPL2B | |
Polyclonal | |
Unconjugated | |
SPPL2B | |
3110056O03Rik; AW550292; IMP4; IMP-4; intramembrane protease 4; KIAA1532; MGC111084; Presenilin homologous protein 4; presenilin-like protein 1; protein SPPL2b; protein SPP-like 2B; PSH4; PSL1; RGD1308556; signal peptide peptidase like 2B; signal peptide peptidase-like 2B; SPPL2B; SPP-like 2B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56928 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TCT7 | |
SPPL2B | |
Synthetic peptide directed towards the N-terminal of human SPPL2B (aa 93-142). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction