Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPPL2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145162
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145162 Supplier Invitrogen™ Supplier No. PA5145162
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HTKGWLAQGQ GAHGLLIVSR VSDQQCSDTT LAPQDPRQPL ADLTIPVAML HYADMLDILS HTRGEAVVRV AMYAPPEPII D Antigen sequence identity: Mouse 67%; Rat 72%.

Intramembrane-cleaving aspartic protease (I-CLiP) that may be able to cleave type II membrane signal peptides in the hydrophobic plane of the membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPPL2C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPPL2C
Gene Accession No. Q8IUH8
Gene Alias IMP5; IMP-5; Intramembrane protease 5; signal peptide peptidase like 2C; signal peptide peptidase-like 2C; SPPL2C; SPP-like 2C
Gene Symbols SPPL2C
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 104-184.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162540
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.