Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPPL2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569941 Supplier Invitrogen™ Supplier No. PA569941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%; Yeast: 75%.

Intramembrane-cleaving aspartic protease (I-CLiP) that may be able to cleave type II membrane signal peptides in the hydrophobic plane of the membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPPL2C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPPL2C
Gene Accession No. Q8IUH8
Gene Alias IMP5; IMP-5; Intramembrane protease 5; signal peptide peptidase like 2C; signal peptide peptidase-like 2C; SPPL2C; SPP-like 2C
Gene Symbols SPPL2C
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human IMP5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162540
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.