Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPR Antibody (OTI4F5), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274344B
Description
SPR Monoclonal specifically detects SPR in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SPR | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SPR | |
Full length recombinant protein of human SPR (NP_003115) produced in HEK293T cells. | |
0.1 mL | |
6697.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4F5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 1.1.1.153, SDR38C1, sepiapterin reductase, sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase), short chain dehydrogenase/reductase family 38C, member 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction