Learn More
Invitrogen™ SPR Polyclonal Antibody

Description
Positive test controls include: HepG2, HeLa, COS-7, Jurkat, MCF7, Mouse liver, Mouse kidney, Mouse pancreas, Rat kidney. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: PLDTDMQQLA RETSVDPDMR KGLQELKAKG KLVDCKVSAQ KLLSLLEKDE FKSGAHVDFY DK.
Specifications
Specifications
| Antigen | SPR |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SPR |
| Gene Accession No. | P18297, P35270, Q64105 |
| Gene Alias | AA409688; ENSMUSG00000071345; Gm10328; SDR38C1; sepiapterin reductase; sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase); short chain dehydrogenase/reductase family 38C, member 1; SPR |
| Gene Symbols | SPR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.