Learn More
Invitrogen™ SPR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588215
Description
Immunogen sequence: MEGGLGRAVC LLTGASRGFG RTLAPLLASL LSPGSVLVLS ARNDEALRQL EAELGAERSG LRVVRVPADL GAEAGLQQLL GALRELPRPK GLQRLLLINN AGSLGDVSKG FVDLSDSTQV NNYWALNLTS MLCLTSSVLK AFPDSPGLNR TVVNISSLCA LQPFKGWALY CAGKAARDML FQVLALEEPN VRVLNYAPGP LDTDMQQLAR ETSVDPDMRK GLQELKAKGK LVDCKVSAQK LLSLLEKDEF KSGAHVDFYD K; Positive Samples: BxPC-3, Jurkat ; Cellular Location: Cytoplasm.
SPR encodes an aldo-keto reductase that catalyzes the NADPH-dependent reduction of pteridine derivatives and is important in the biosynthesis of tetrahydrobiopterin (BH4). Mutations in this gene result in DOPA-responsive dystonia due to sepiaterin reductase deficiency. A pseudogene has been identified on chromosome 1.
Specifications
SPR | |
Polyclonal | |
Unconjugated | |
SPR | |
AA409688; ENSMUSG00000071345; Gm10328; SDR38C1; sepiapterin reductase; sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase); short chain dehydrogenase/reductase family 38C, member 1; SPR | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6697 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.96 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P35270 | |
SPR | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-261 of human SPR (NP_0031151). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.