Learn More
Description
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.5) containing 1mM DTT, 10% glycerol containingno Preservative |
| Gene ID (Entrez) | 6697 |
| Name | Recombinant SPR Protein |
| Purification Method | >95%, by SDS-PAGE |
| Quantity | 500 μg |
| Source | E. coli |
| Storage Requirements | Store at 4°Cshort term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
